missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4-dichloropyrimidine, TRC

Product Code. 30311012
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
This item is not returnable. View return policy

Product Code. 30311012

Brand: Toronto Research Chemicals TRCA6047202.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Amino-2,4-dichloropyrimidine
CAS 5177-27-5
Molecular Formula C4H3Cl2N3
Quantity 2.5 g
Synonym 2,4-Dichloro-5-pyrimidinamine, NSC 86381
InChI Formula InChI=1S/C4H3Cl2N3/c5-3-2(7)1-8-4(6)9-3/h1H,7H2
SMILES Nc1cnc(Cl)nc1Cl
IUPAC Name 2,4-dichloropyrimidin-5-amine
Molecular Weight (g/mol) 163.99
Formula Weight 162.9704
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.