missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-mercaptobenzimidazole, 96%

Product Code. 11491640
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11491640 100 g 100g
11481640 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11491640

Brand: Thermo Scientific Alfa Aesar H26999.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2818-66-8 | C7H7N3S | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2818-66-8
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00022671
InChI Key BXDMTLVCACMNJO-UHFFFAOYSA-N
Synonym 5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol
PubChem CID 2734002
IUPAC Name 5-amino-1,3-dihydrobenzimidazole-2-thione
SMILES NC1=CC=C2NC(=S)NC2=C1

Specifications

Melting Point 240°C to 244°C
Quantity 100 g
Formula Weight 165.22
Percent Purity 96%
Chemical Name or Material 5-Amino-2-mercaptobenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.