missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-methoxypyridine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 5-Amino-2-methoxypyridine |
| CAS | 6628-77-9 |
| Molecular Formula | C6H8N2O |
| Quantity | 5 g |
| Synonym | (2-Methoxypyridin-5-yl)amine, 2-Methoxy-5-aminopyridine, 3-Amino-6-methoxypyridine, 6-Methoxy-3-aminopyridine, 6-Methoxy-3-pyridinamine, 6-Methoxy-3-pyridinylamine, NSC 59708 |
| InChI Formula | InChI=1S/C6H8N2O/c1-9-6-3-2-5(7)4-8-6/h2-4H,7H2,1H3 |
| SMILES | COc1ccc(N)cn1 |
| IUPAC Name | 6-methoxypyridin-3-amine |
| Molecular Weight (g/mol) | 124.14 |
| Formula Weight | 124.0637 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction