missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-naphthalenesulfonic Acid, ~90%, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 5-Amino-2-naphthalenesulfonic Acid |
| CAS | 119-79-9 |
| Molecular Formula | C10H9NO3S |
| Quantity | 100 g |
| Synonym | 1-Naphthylamine-6-Sulfonic Acid, Cleve's beta-Acid, 6-Sulfo-1-naphthylamine, NSC 31506, NSC 4984 |
| InChI Formula | InChI=1S/C10H9NO3S/c11-10-3-1-2-7-6-8(15(12,13)14)4-5-9(7)10/h1-6H,11H2,(H,12,13,14) |
| SMILES | Nc1cccc2cc(ccc12)S(=O)(=O)O |
| IUPAC Name | 5-aminonaphthalene-2-sulfonic acid |
| Molecular Weight (g/mol) | 223.25 |
| Formula Weight | 223.0303 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction