missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(trifluoromethyl)pyridine, 98%

Product Code. 15453518
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453518 1 g 1g
15473518 250 mg 250mg
15463518 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15453518 Supplier Thermo Scientific Alfa Aesar Supplier No. H56968.03
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106877-33-2 | C6H5F3N2 | 162.12 g/mol

Employed in a convenient, one-pot, synthesis of azaindoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in a convenient, one-pot, synthesis of azaindoles.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106877-33-2
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.12
MDL Number MFCD01862656
InChI Key IBOZOWZSXZNIHI-UHFFFAOYSA-N
Synonym 5-amino-2-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-3-amine, 3-amino-6-trifluoromethyl pyridine, 5-amino-2-trifluoromethylpyridine, 3-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-3-pyridinamine, 3-pyridinamine, 6-trifluoromethyl, 6-trifluoromethyl-3-pyridylamine, 2-trifluoromethyl-5-aminopyridine, pubchem2997
PubChem CID 2737744
IUPAC Name 6-(trifluoromethyl)pyridin-3-amine
SMILES NC1=CC=C(N=C1)C(F)(F)F

Specifications

Melting Point 39°C to 43°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 162.11
Percent Purity 98%
Chemical Name or Material 5-Amino-2-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.