missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole, 96%

Product Code. p-7032118
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448485 250 mg 250mg
15436277 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448485 Supplier Thermo Scientific Alfa Aesar Supplier No. H33467.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19922-07-7 | C8H6ClN3S | 211.667 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19922-07-7
Molecular Formula C8H6ClN3S
Molecular Weight (g/mol) 211.667
MDL Number MFCD07357326
InChI Key IUSHKZWNRIGLGO-UHFFFAOYSA-N
Synonym 3-4-chlorophenyl-1,2,4-thiadiazol-5-amine, 1,2,4-thiadiazol-5-amine,3-4-chlorophenyl, 5-amino-3-4-chlorophenyl-1,2,4-thiadiazole, chlorophenylthiadiazolamine, 3-4-chlorophenyl-1,2,4-thiadiazole-5-ylamine
PubChem CID 23331270
IUPAC Name 3-(4-chlorophenyl)-1,2,4-thiadiazol-5-amine
SMILES C1=CC(=CC=C1C2=NSC(=N2)N)Cl

Specifications

Melting Point 183°C to 185°C
Quantity 250 mg
Formula Weight 211.67
Percent Purity 96%
Chemical Name or Material 5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.