missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(4-methylphenyl)pyrazole, 97%, Thermo Scientific™

Product Code. 11996211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11996211 5 g 5g
11986211 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11996211 Supplier Thermo Scientific Acros Supplier No. 438800050
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 78597-54-3
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
InChI Key GVPFRVKDBZWRCZ-UHFFFAOYSA-N
Synonym 5-p-tolyl-1h-pyrazol-3-amine, 3-4-methylphenyl-1h-pyrazol-5-amine, 5-p-tolyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methylphenyl pyrazole, 3-amino-5-4-methylphenyl-1h-pyrazole, 3-amino-5-p-tolylpyrazole, 5-4-methylphenyl-1h-pyrazol-3-amine, 3-p-tolyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-4-methylphenyl, 5-4-methylphenyl-2h-pyrazol-3-amine
PubChem CID 736768
IUPAC Name 5-(4-methylphenyl)-1H-pyrazol-3-amine
SMILES CC1=CC=C(C=C1)C2=CC(=NN2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.3°C to 153.3°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 173.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Amino-3-(4-methylphenyl)pyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-3-(4-methylphenyl)pyrazole, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.