missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-methyl-1-phenyl-1H-pyrazole, ≥99%, Thermo Scientific Chemicals

Product Code. 15420389
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420389 5 g 5g
15430389 25 g 25g
15440389 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420389 Supplier Thermo Scientific Alfa Aesar Supplier No. H63605.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1131-18-6 | Molecular Formula: C10H11N3 | Molecular Weight: 173.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-18-6
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
MDL Number MFCD00020727
InChI Key FMKMKBLHMONXJM-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl
PubChem CID 70801
IUPAC Name 5-methyl-2-phenylpyrazol-3-amine
SMILES CC1=NN(C(=C1)N)C2=CC=CC=C2

Specifications

Melting Point 114°C to 117°C
Color Brown to Yellow
Quantity 5 g
Beilstein 140666
Formula Weight 173.21
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 5-Amino-3-methyl-1-phenyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.