missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4-(4-fluorophenyl)-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 15475597
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475597 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15475597 Supplier Thermo Scientific Alfa Aesar Supplier No. H32831.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1442104
Melting Point 160°C to 162°C
Molecular Formula CBi2O5
MDL Number MFCD03001202
Quantity 1 g
Synonym bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
SMILES C(=O)(O[Bi]=O)O[Bi]=O
IUPAC Name bis(oxobismuthanyl) carbonate
Molecular Weight (g/mol) 509.97
PubChem CID 16683095
ChEBI CHEBI:31291
Formula Weight 177.18
Percent Purity 0.97
Chemical Name or Material 5-Amino-4-(4-fluorophenyl)-1H-pyrazole
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.