missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4,6-dichloropyrimidine, 98+%

Product Code. 11411567
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411567 5 g 5g
11401567 1 g 1g
11421567 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411567 Supplier Thermo Scientific Alfa Aesar Supplier No. B24985.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5413-85-4 | C4H3Cl2N3 | 163.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5413-85-4
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.99
MDL Number MFCD00006108
InChI Key NIGDWBHWHVHOAD-UHFFFAOYSA-N
Synonym 5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine
PubChem CID 79434
IUPAC Name 4,6-dichloropyrimidin-5-amine
SMILES NC1=C(Cl)N=CN=C1Cl

Specifications

Melting Point 143°C to 146°C
Odor Irritating
Quantity 5 g
Beilstein 126885
Sensitivity Moisture sensitive
Formula Weight 164
Percent Purity ≥98%
Chemical Name or Material 5-Amino-4,6-dichloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.