missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminoimidazole-4-carboxamide-1-Beta-D-ribofuranoside, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside |
| CAS | 2627-69-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 H14 N4 O5 |
| Quantity | 1 g |
| Synonym | 1-Ribosyl-4-carboxamido-5-aminoimidazole, 5-Amino-1-ribosyl-4-imidazolecarboxamide, 5-Amino-4-imidazolecarboxamide ribofuranoside, AIC-Riboside, AICA-Riboside, AICAR, Acadesine, Arasine, GP 1-110, NSC 105823 |
| InChI Formula | InChI=1S/C9H14N4O5/c10-7-4(8(11)17)12-2-13(7)9-6(16)5(15)3(1-14)18-9/h2-3,5-6,9,14-16H,1,10H2,(H2,11,17)/t3-,5-,6-,9-/m1/s1 |
| SMILES | NC(=O)c1ncn([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c1N |
| IUPAC Name | 5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]imidazole-4-carboxamide |
| Molecular Weight (g/mol) | 258.23 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction