missing translation for 'onlineSavingsMsg'
Learn More

Melford 5-Aminolevulinic Acid

Product Code. 30017943
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017943 5 g 5g
30017942 1 g 1g
30017941 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30017943 Supplier Melford Supplier No. A112505.0
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Substrate for detection of delta-aminolevulinic acid dehydratase.

Substrate for detection of delta-aminolevulinic acid dehydratase.

Specifications

CAS 9-2-5451
Molecular Formula C5H9NO3-HCl
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.