missing translation for 'onlineSavingsMsg'
Learn More

5-Aminolevulinic acid hydrochloride, 99%

Product Code. 11314529
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
Unit Size:
100mg
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314529 2 g 2g
11488776 100 mg 100mg
11498776 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314529 Supplier Thermo Scientific Alfa Aesar Supplier No. A16942.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5451-09-2 | C5H10ClNO3 | 167.59 g/mol

5-Aminolevulinic acid hydrochloride finds an important role as a precursor in the synthesis of tetrapyrroles such as chlorophyll and heme. It is widely utilized in photodynamic therapy of diseases namely, Paget’s disease and human papillomavirus (HPV) infection-associated cervical condylomata acuminata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminolevulinic acid hydrochloride finds an important role as a precursor in the synthesis of tetrapyrroles such as chlorophyll and heme. It is widely utilized in photodynamic therapy of diseases namely, Paget’s disease and human papillomavirus (HPV) infection-associated cervical condylomata acuminata.

Solubility
Soluble in dimethyl sulfoxide, methanol and water.

Notes
Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9-2-5451
Molecular Formula C5H10ClNO3
Molecular Weight (g/mol) 167.59
MDL Number MFCD00012869
InChI Key ZLHFONARZHCSET-UHFFFAOYSA-N
Synonym 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
PubChem CID 123608
IUPAC Name 5-amino-4-oxopentanoic acid;hydrochloride
SMILES C(CC(=O)O)C(=O)CN.Cl

Specifications

Melting Point ∼156°C (decomposition)
Beilstein 3690651
Merck Index 14,446
Solubility Information Soluble in dimethyl sulfoxide,methanol and water.
Quantity 2 g
Formula Weight 167.59
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material 5-Aminolevulinic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.