missing translation for 'onlineSavingsMsg'
Learn More

5-Aminolevulinic acid hydrochloride, MedChemExpress

Product Code. 16451378
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
10 g
Unit Size:
100mg
10g
1g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451378 5 g 5g
16442519 1 g 1g
16409056 100 mg 100mg
16428948 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16451378 Supplier Medchem Express Supplier No. HYN0305
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 5-Aminolevulinic acid hydrochloride (5-ALA hydrochloride) is an intermediate in heme biosynthesis in the body and the universal precursor of tetrapyrroles.

5-Aminolevulinic acid hydrochloride (5-ALA hydrochloride) is an intermediate in heme biosynthesis in the body and the universal precursor of tetrapyrroles.

Specifications

Chemical Name or Material 5-Aminolevulinic acid hydrochloride
CAS 9-2-5451
Color White
Purity Grade Notes Research
Molecular Formula C5H10ClNO3
Quantity 5 g
Synonym 5-ALA hydrochlorideδ-Aminolevulinic acid hydrochloride5-Amino-4-oxopentanoic acid hydrochloride
Solubility Information DMSO : 100 mg/mL (596.69 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (99.47 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CCC(CN)=O.[H]Cl
Molecular Weight (g/mol) 167.59
Formula Weight 167.59
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.