missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ 5-(Aminomethyl)Fluorescein, Hydrochloride

Product Code. 11553997
Change view
Click to view available options
Quantity:
10 mg
Packaging:
1 Each
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11553997 10 mg 1 Each 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11553997 Supplier Molecular Probes™ Supplier No. A1353

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-(aminomethyl)fluorescein, hydrochloride

The primary aliphatic amine of 5-(aminomethyl)fluorescein can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
TRUSTED_SUSTAINABILITY

Specifications

CAS 138588-53-1
Color Orange-Yellow
Packaging 1 Each
Molecular Formula C21H15NO5
Quantity 10 mg
Synonym 5-aminomethyl fluorescein, 5-aminomethyl fluorescein hydrochloride, 5-amfl, 6-aminomethyl-3',6'-dihydroxyspiro 2-benzofuran-3,9'-xanthene-1-one, 5'-aminomethyl-3,6-dihydroxyspiro 9h-xanthene-9,1' 3'h-isobenzofuran-3'-one, 5-aminomethyl-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one
InChI Key FKQRDXNGJULUOJ-UHFFFAOYSA-N
Molecular Weight (g/mol) 361.35
PubChem CID 132166

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.