missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Aminonicotinic acid, 97%

Product Code. 15456668
Change view
Click to view available options
Quantity:
5g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456668 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456668 Supplier Alfa Aesar™ Supplier No. H60192.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 24242-19-1
Molecular Formula C6H6N2O2
MDL Number MFCD00129116
Quantity 5g
Beilstein 115848
Synonym 5-aminonicotinic acid, 5-amino-nicotinic acid, 5-aminonicotinicacid, 3-pyridinecarboxylic acid, 5-amino, 5-amino nicotinic acid, 5-amino-3-carboxypyridine, 5-amino-3-pyridinecarboxylic acid, zlchem 307, pubchem15914, acmc-209ga2
InChI Key BYIORJAACCWFPU-UHFFFAOYSA-N
SMILES C1=C(C=NC=C1N)C(=O)O
IUPAC Name 5-aminopyridine-3-carboxylic acid
Molecular Weight (g/mol) 138.126
PubChem CID 354316
ChEBI CHEBI:68578
Formula Weight 138.13
Percent Purity 97%
Chemical Name or Material 5-Aminonicotinic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.