missing translation for 'onlineSavingsMsg'
Learn More

5-Aza-2'-deoxycytidine, 98%

Product Code. 15436949
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436949 250 mg 250mg
15426949 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436949 Supplier Thermo Scientific Alfa Aesar Supplier No. J65513.MD
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2353-33-5 | C9H13N3O4 | 227.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-33-5
Molecular Formula C9H13N3O4
Molecular Weight (g/mol) 227.22
MDL Number MFCD00006547
InChI Key CKTSBUTUHBMZGZ-SHYZEUOFSA-N
Synonym decitabine, 5-aza-2'-deoxycytidine, dacogen, 5-azadeoxycytidine, 2'-deoxy-5-azacytidine, azadc, dezocitidine, 5-aza-cdr, 5-aza-dc, dacogen tn
PubChem CID 451668
ChEBI CHEBI:50131
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Color White
Quantity 250 mg
Beilstein 617982
Merck Index 14,2853
Formula Weight 228.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Aza-2'-deoxycytidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.