missing translation for 'onlineSavingsMsg'
Learn More

5-Azabenzimidazole, 98%, Thermo Scientific Chemicals

Product Code. 11317056
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317056 1 g 1g
11327056 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317056 Supplier Thermo Scientific Alfa Aesar Supplier No. L06930.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

5-Azabenzimidazole is used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Azabenzimidazole is used as Pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 272-97-9
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD00051808
InChI Key UBOOKRVGOBKDMM-UHFFFAOYSA-N
Synonym 3h-imidazo 4,5-c pyridine, 1h-imidazo 4,5-c pyridine, 5-azabenzimidazole, 3,5-diazaindole, 5h-imidazo 4,5-c pyridine, imidazo 4,5-c pyridine, 5-aza-1h-benzoimidazole, 5-aza-benzimidazole
PubChem CID 9227
IUPAC Name 3H-imidazo[4,5-c]pyridine
SMILES N1C=NC2=CC=NC=C12

Specifications

Melting Point 170°C to 173°C
Quantity 1 g
Beilstein 2601
Formula Weight 119.13
Percent Purity 98%
Chemical Name or Material 5-Azabenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.