missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies 5-Azacytidine
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
Description
- 5-Azacytidine is an analog of the nucleoside cytidine which can be incorporated into DNA and RNA
- 5-Azacytidine acts as an epigenetic modifier by incorporating into DNA where it irreversibly binds to DNA methyltransferases, thus inhibiting their activity
- Increases reprogramming efficiency of mouse fibroblasts to induced pluripotent stem (iPS) cells by inducing full reprogramming of partially reprogrammed cells (Mikkelsen et al.)
- Resets epigenetic memory in mouse iPS cells, in combination with Trichostatin A (Kim et al.)
- Enhances differentiation to cardiomyocytes from human embryonic stem cells (Yoon et al.)
- Wide range of anti-metabolic activities when tested against cultured cancer cells and an effective chemotherapeutic agent for acute myelogenous leukemia
Specifications
Specifications
| CAS | 320-67-2 |
| Synonym | 5-AzaC; Antibiotic U 18496; Ladakamycin; Mylosar; NSC 103-627; NSC 102816; U 18496; WR 183027; Zcyd |
| Quantity | 250 mg |
| Molecular Formula | C8H12N4O5 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction