Learn More
5-Azacytidine, 98%, Thermo Scientific™
Causes DNA demethylation or hemi-demethylation; a powerful mutagen
Brand: Thermo Scientific Alfa Aesar J64159.06
| Quantity | 5g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications5-Azacytidine, its incorporation into RNA alters RNA synthesis and processing, and results in inhibition of protein synthesis. It has been used as a cancer chemotherapeutic agent. It is a powerful bacteriostatic, antitumor, and mutagenic agent; it also exhibits immunosuppressive, antimitotic, radioprotective, and virostatic effects.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
| 320-67-2 | |
| 244.207 | |
| NMUSYJAQQFHJEW-KVTDHHQDSA-N | |
| 9444 | |
| 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one |
| C8H12N4O5 | |
| MFCD00006539 | |
| 5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin | |
| CHEBI:2038 | |
| C1=NC(=NC(=O)N1C2C(C(C(O2)CO)O)O)N |
Specifications
| 320-67-2 | |
| MFCD00006539 | |
| 620461 | |
| 5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin | |
| NMUSYJAQQFHJEW-KVTDHHQDSA-N | |
| 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one | |
| 9444 | |
| 244.2 | |
| 5-Azacytidine |
| C8H12N4O5 | |
| 5g | |
| 14,890 | |
| Soluble in water. | |
| C1=NC(=NC(=O)N1C2C(C(C(O2)CO)O)O)N | |
| 244.207 | |
| CHEBI:2038 | |
| 98% |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.