missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 5-Azacytidine
GREENER_CHOICE

Product Code. 18083992
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18083992

Brand: Tocris Bioscience™ 3842/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DNA methyltransferase inhibitor

DNA methyltransferase inhibitor. Incorporates into DNA forming covalent adducts with cellular DNMT1, depleting enzyme activity. Induces demethylation and reactivation of silenced genes. Improves the efficiency of reprogramming of stem cells; induces differentiation of mesenchymal stem cells into cardiomyocytes.
TRUSTED_SUSTAINABILITY

Specifications

CAS 320-67-2
Quantity 50 mg
Molecular Formula C8H12N4O5
Formula Weight 244.2
Purity >98%
Molecular Weight (g/mol) 244.21
Chemical Name or Material 5-Azacytidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.