missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Azacytidine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 5-Azacytidine |
| CAS | 320-67-2 |
| Molecular Formula | C8 H12 N4 O5 |
| Quantity | 250 mg |
| Synonym | 4-Amino-1-β-D-ribofuranosyl-1,3,5-triazin-2(1H)-one, s-Triazin-2(1H)-one, 4-amino-1-β-D-ribofuranosyl- (8CI), 5-AC, 5-AZC, 5-AZCR, 5-AzaC, 5-Azacytidine, Antibiotic U 18496, Azacitidine, Azacytidine, Ladakamycin, Ledakamycin, Mylosar, NSC 102816, NSC 103-627, U 18496, Vidaza, WR 183027 |
| InChI Formula | InChI=1S/C8H12N4O5/c9-7-10-2-12(8(16)11-7)6-5(15)4(14)3(1-13)17-6/h2-6,13-15H,1H2,(H2,9,11,16)/t3-,4-,5-,6-/m1/s1 |
| SMILES | NC1=NC(=O)N(C=N1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O |
| IUPAC Name | 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one |
| Molecular Weight (g/mol) | 244.2 |
| Formula Weight | 244.0808 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction