missing translation for 'onlineSavingsMsg'
Learn More

5-[(Benzoylamino)methyl]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10242642
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10242642 1 g Amber Glass Bottle 1g
10160972 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10242642 Supplier Thermo Scientific Maybridge Supplier No. KM04426DA
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 138872-44-3
Molecular Formula C12H10ClNO3S2
Molecular Weight (g/mol) 315.786
MDL Number MFCD00052130
InChI Key VGSWVDWOXYTAPG-UHFFFAOYSA-N
Synonym 5-benzoylamino methyl thiophene-2-sulfonyl chloride, 5-benzamidomethyl thiophene-2-sulfonyl chloride, 2-benzoylaminomethyl thiophene-5-sulfonylchloride, 2-benzoylaminomethyl thiophene-5-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-benzoylamino methyl, 5-phenylformamido methyl thiophene-2-sulfonyl chloride, 5-benzoylamino methyl thiophene-2-sulphonyl chloride, pubchem5470, acmc-20c3wu, 5-benzamidomethyl thiophene-2-sulfonylchloride
PubChem CID 2778878
IUPAC Name 5-(benzamidomethyl)thiophene-2-sulfonyl chloride
SMILES C1=CC=C(C=C1)C(=O)NCC2=CC=C(S2)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 315.8
Percent Purity 97%
Chemical Name or Material 5-[(Benzoylamino)methyl]thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.