missing translation for 'onlineSavingsMsg'
Learn More

5-(Benzyloxycarbonylamino)valeric acid, 97%

Product Code. 15472389
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15472389 5 g 5g
15482389 250 mg 250mg
15462389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15472389 Supplier Thermo Scientific Alfa Aesar Supplier No. H64958.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 23135-50-4 | C13H17NO4 | 251.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23135-50-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00188458
InChI Key QYYPKLYDFCYGPG-UHFFFAOYSA-N
Synonym 5-benzyloxy carbonyl amino pentanoic acid, 5-benzyloxycarbonylamino-pentanoic acid, 5-benzyloxycarbonylamino pentanoic acid, z-5-ava-oh, 5-carbobenzoxyamino valeric acid, 5-carbobenzoxyamino pentanoic acid, 5-phenylmethoxycarbonylamino pentanoic acid, 5-benzyloxycarbonylamino-pentanoicacid, pubchem20738
PubChem CID 3690024
SMILES OC(=O)CCCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 99°C to 105°C
Color White
Linear Formula C6H4CH2OCONH2(CH2)4CO2H
Beilstein 2334766
IUPAC Name 5-(phenylmethoxycarbonylamino)pentanoic acid
Quantity 5 g
Formula Weight 251.28
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-(Benzyloxycarbonylamino)valeric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.