missing translation for 'onlineSavingsMsg'
Learn More

5-BOC-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine, 97%, Thermo Scientific™

Product Code. 15522313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15522313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15522313 Supplier Thermo Scientific Acros Supplier No. 450020010
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 230301-11-8
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C11H17N3O2
Quantity 1 g
Synonym tert-butyl 6,7-dihydro-1h-pyrazolo 4,3-c pyridine-5 4h-carboxylate, 5-boc-1,4,6,7-tetrahydropyrazolo 4,3-c pyridine, tert-butyl 6,7-dihydro-2h-pyrazolo 4,3-c pyridine-5 4h-carboxylate, 5-boc-4,5,6,7-tetrahydropyrazolo 4,3-c pyridine, tert-butyl 2h,4h,5h,6h,7h-pyrazolo 4,3-c pyridine-5-carboxylate, tert-butyl 1h,4h,5h,6h,7h-pyrazolo 4,3-c pyridine-5-carboxylate, tert-butyl 2,4,6,7-tetrahydro-5h-pyrazolo-4,3-c pyridine-5-carboxylate, tert-butyl 2,4,6,7-tetrahydro-5h-pyrazolo 4,3-c pyridine-5-carboxylate, t-butyl 1h,4h,6h,7h-pyrazolo 4,3-c pyridine-5-carboxylate, tert-butyl 1h,4h,6h,7h-pyrazolo 4,3-c pyridine-5-carboxylate
InChI Key BHYPERDTLBCHAE-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCC2=C(C1)C=NN2
IUPAC Name tert-butyl 1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine-5-carboxylate
Molecular Weight (g/mol) 223.27
PubChem CID 10398680
Formula Weight 223.27
Percent Purity 97%
Chemical Name or Material 5-BOC-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-BOC-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.