missing translation for 'onlineSavingsMsg'
Learn More

5-(Boc-amino)-1-pentanol, 96%

Product Code. 11378586
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378586 5 g 5g
11368586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378586 Supplier Thermo Scientific Alfa Aesar Supplier No. L17649.06
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 75178-90-4 | C10H21NO3 | 203.28 g/mol

5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.

Solubility
Difficult to mix in water.

Notes
Moisture sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75178-90-4
Molecular Formula C10H21NO3
Molecular Weight (g/mol) 203.28
MDL Number MFCD01862955
InChI Key DDGNGFVNTZJMMZ-UHFFFAOYSA-N
Synonym 5-boc-amino-1-pentanol, tert-butyl n-5-hydroxypentyl carbamate, tert-butyl 5-hydroxypentyl carbamate, 5-tert-butoxycarbonylamino-1-pentanol, n-5-hydroxypentyl carbamic acid tert-butyl ester, tert-butyl 5-hydroxypentylcarbamate, carbamic acid, 5-hydroxypentyl-, 1,1-dimethylethyl ester, n-boc-5-aminopentanol, 5-boc-amino 1-pentanol, acmc-209ox9
PubChem CID 4456136
IUPAC Name tert-butyl N-(5-hydroxypentyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCO

Specifications

Linear Formula (CH3)3COCONH(CH2)5OH
Refractive Index 1.456
Beilstein 3603659
Solubility Information Difficult to mix in water.
Density 1
Quantity 5 g
Flash Point >109°C (228°F)
Formula Weight 203.28
Percent Purity 96%
Chemical Name or Material 5-(Boc-amino)-1-pentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.