missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5-BrdU

Product Code. 16441796
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441796 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441796 Supplier MedChemExpress Supplier No. HY15910
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5-BrdU (BrdU) is a nucleoside analog that competes with thymidine for incorporation into DNA. 5-BrdU is commonly used in the detection of proliferating cells.

Specifications

Molecular Weight (g/mol) 307.1 g/mol
Product Type Reference compound
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Shipping Condition Room temperature
Purity 95.0%
Quantity 500 mg
Solubility Information DMSO : 250 mg/mL (814.07 mM∣Need ultrasonic) ∣H2O : 2 mg/mL (6.51 mM∣ultrasonic and warming and heat to 60°C)
For Use With (Application) Cancer-programmed cell death
CAS 59-14-3
Synonym BrdU∣ 5-Bromo-2'-deoxyuridine∣ BUdR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.