missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,3,4-thiadiazol-2-ylamine, 97%, Thermo Scientific™

Product Code. 10225504
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10225504 1 g 1g
10663425 5 g 5g
10193354 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10225504 Supplier Thermo Scientific Maybridge Supplier No. MO08007DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 37566-39-5
Molecular Formula C2H2BrN3S
Molecular Weight (g/mol) 180.02
MDL Number MFCD00464325
InChI Key GLYQQFBHCFPEEU-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-1,3,4-thiadiazole, 5-bromo-1,3,4-thiadiazol-2-ylamine, 2-amino-5-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazol-2-amine, 5-bromo, 5-bromo-1,3,4-thiadiazole-2-amine, 5-bromo-1,3,4-thiadiazole-2-ylamine, 5-bromo-1,3,4 thiadiazol-2-ylamine, bromothiadiazolamine, pubchem20972, ksc496o1t
PubChem CID 3650883
IUPAC Name 5-bromo-1,3,4-thiadiazol-2-amine
SMILES NC1=NN=C(Br)S1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 180.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1, 3, 4-thiadiazol-2-ylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.