missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,4-benzodioxane, 95%

Product Code. 15463757
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463757 250 mg 250mg
11382819 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463757 Supplier Thermo Scientific Alfa Aesar Supplier No. H27709.MD
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58328-39-5 | C8H7BrO2 | 215.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58328-39-5
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD03095029
InChI Key NGOZRIZXELGVHK-UHFFFAOYSA-N
Synonym 5-bromo-1,4-benzodioxane, 5-bromo-2,3-dihydro-1,4-benzodioxane, 5-bromo-2,3-dihydrobenzo b 1,4 dioxine, acmc-209m3q, 5-bromo-2,3-dihydrobenzo 1,4 dioxin, 5-bromo-2,3-dihydro 1,4 benzodioxine, 5-bromo-2,3-dihydro 1,4-benzodioxine, 1,4-benzodioxin, 5-bromo-2,3-dihydro
PubChem CID 18787268
IUPAC Name 5-bromo-2,3-dihydro-1,4-benzodioxine
SMILES C1COC2=C(O1)C=CC=C2Br

Specifications

Refractive Index 1.5955
Quantity 250 mg
Formula Weight 215.05
Percent Purity 95%
Chemical Name or Material 5-Bromo-1,4-benzodioxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.