missing translation for 'onlineSavingsMsg'
Learn More

(5-Bromo-1-benzofuran-2-yl)methanol, ≥97%, Thermo Scientific™

Product Code. 10624405
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10624405 5 g 5g
10026292 250 mg 250mg
10321143 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10624405 Supplier Thermo Scientific Maybridge Supplier No. CC31209DE
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 38220-77-8
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD06659073
InChI Key JYYWIDBNICYLBN-UHFFFAOYSA-N
Synonym 5-bromo-1-benzofuran-2-yl methanol, 5-bromobenzofuran-2-yl methanol, 5-bromobenzo d furan-2-yl methan-1-ol, 5-bromo-2-benzofuranylmethanol, 5-bromo-2-benzofuranyl methanol, 5-bromo-benzofuran-2-yl-methanol, 5-bromo-2-hydroxymethyl-1-benzofuran, 5-bromo-1-benzo b furan-2-yl methanol
PubChem CID 2795537
IUPAC Name (5-bromo-1-benzofuran-2-yl)methanol
SMILES OCC1=CC2=CC(Br)=CC=C2O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 227.06
Percent Purity 97%
Chemical Name or Material (5-Bromo-1-benzofuran-2-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.