missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-benzothiophene, 97%, Thermo Scientific™

Product Code. 10069431
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10069431 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10069431 Supplier Thermo Scientific Maybridge Supplier No. CC24810DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4923-87-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H5BrS
MDL Number MFCD03069318
Quantity 1g
Synonym 5-bromobenzo b thiophene, 5-bromobenzothiophene, 5-bromo-benzo b thiophene, 5-bromothianaphthene, benzo b thiophene, 5-bromo, zlchem 181, pubchem7752, 5-bromo-benzothiophene, pubchem15353, acmc-1anox
InChI Key RDSIMGKJEYNNLF-UHFFFAOYSA-N
SMILES BrC1=CC=C2SC=CC2=C1
IUPAC Name 5-bromo-1-benzothiophene
Molecular Weight (g/mol) 213.09
PubChem CID 2776578
Formula Weight 213.1
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-benzothiophene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.