missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%

Product Code. 10734312 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10734312 5 g Glass bottle 5g
10342020 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10734312 Supplier Thermo Scientific Acros Supplier No. 431170050
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 400071-95-6 | C10H8BrNO2 | 254.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400071-95-6
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.08
MDL Number MFCD14706162
InChI Key WQZLFFJMRKTCMU-UHFFFAOYSA-N
Synonym 5-bromo-1-methyl-1h-indole-3-carboxylic acid, 5-bromo-1-methyl-1h-indole-3-carboxylicacid, 1h-indole-3-carboxylic acid, 5-bromo-1-methyl, ksc230c9b, 5-bromo-1-methyl-1h-indole-3-carboxylic acid;
PubChem CID 11032413
IUPAC Name 5-bromo-1-methylindole-3-carboxylic acid
SMILES CN1C=C(C2=C1C=CC(=C2)Br)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 255.4°C to 255.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 254.08
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1-methyl-1H-indole-3-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.