missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methylindoline, 90%, Thermo Scientific™

Product Code. 10356374
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356374 250 mg Amber Glass Bottle 250mg
10517284 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10356374 Supplier Thermo Scientific Maybridge Supplier No. CC58710CB
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 99848-78-9
Molecular Formula C9H10BrN
Molecular Weight (g/mol) 212.09
MDL Number MFCD09817498
InChI Key YIRSVRZXIXHOSN-UHFFFAOYSA-N
Synonym 5-bromo-1-methylindoline, 5-bromo-1-methyl-2,3-dihydro-1h-indole, 1h-indole, 5-bromo-2,3-dihydro-1-methyl, 1-methyl-5-bromo-2,3-dihydro-1h-indole, 5-bromo-2,3-dihydro-1-methyl-1h-indole
PubChem CID 19361435
SMILES CN1CCC2=C1C=CC(Br)=C2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 5-bromo-1-methyl-2,3-dihydroindole
Formula Weight 212.09
Percent Purity 90%
Chemical Name or Material 5-Bromo-1-methylindoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.