missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-pentene, 96%

Product Code. p-7081258
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394669 1 g 1g
11443544 5 g 5g
11453544 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394669 Supplier Thermo Scientific Alfa Aesar Supplier No. A11219.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1119-51-3 | C5H9Br | 149.031 g/mol

5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.

Solubility
Immiscible with water.

Notes
Keep away from sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-51-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD0000-364
InChI Key LPNANKDXVBMDKE-UHFFFAOYSA-N
Synonym 5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide
PubChem CID 70704
IUPAC Name 5-bromopent-1-ene
SMILES C=CCCCBr

Specifications

Density 1.259
Boiling Point 126°C to 127°C
Flash Point 30°C (86°F)
Linear Formula Br(CH2)3CH=CH2
Refractive Index 1.4639
Quantity 1 g
UN Number UN1993
Beilstein 506077
Solubility Information Immiscible with water.
Formula Weight 149.03
Percent Purity 96%
Chemical Name or Material 5-Bromo-1-pentene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.