missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-benzimidazole, 97%, Thermo Scientific™

Product Code. 10505004
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10505004 10 g Amber Glass Bottle 10g
10039602 1 g Amber Glass Bottle 1g
10472854 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10505004 Supplier Thermo Scientific Maybridge Supplier No. CC45910EA
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4887-88-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD00160001
InChI Key GEDVWGDBMPJNEV-UHFFFAOYSA-N
Synonym 5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
PubChem CID 785299
SMILES BrC1=CC=C2N=CNC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name 6-bromo-1H-benzimidazole
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1H-benzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.