missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-benzimidazole, 97%

Product Code. 15274884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15274884 25 g 25g
15254884 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15274884 Supplier Thermo Scientific Acros Supplier No. 451670250
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4887-88-1 | C7H5BrN2 | 197.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4887-88-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD00160001
InChI Key GEDVWGDBMPJNEV-UHFFFAOYSA-N
Synonym 5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
PubChem CID 785299
SMILES BrC1=CC=C2N=CNC2=C1

Specifications

Melting Point 130.0°C to 134.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 25 g
IUPAC Name 6-bromo-1H-1,3-benzodiazole
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1H-benzimidazole
compliance-icons
Product Identifier
  • 5-Bromo-1H-benzimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.