missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,2-difluoro-1,3-benzodioxole, 97%

Product Code. 11440068
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440068 5 g 5g
11430068 1 g 1g
11384669 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440068 Supplier Thermo Scientific Alfa Aesar Supplier No. B23661.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33070-32-5 | C7H3BrF2O2 | 237 g/mol

5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.

Solubility
Miscible with hexane.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33070-32-5
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237
MDL Number MFCD00236212
InChI Key SZRHWHHXVXSGMT-UHFFFAOYSA-N
Synonym 5-bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2h-1,3-benzodioxole, 5-bromo-2,2-difluorobenzo d 1,3 dioxole, 5-bromo-2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 5-bromo-2,2-difluoro, 4-bromo-1,2-difluoromethylenedioxy benzene, 5-bromo-2,2-difluorobenzo d 1,3-dioxolene, acmc-209hy8, 5-bromo2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-5-bromo-1,3-benzodioxole
PubChem CID 2736271
IUPAC Name 5-bromo-2,2-difluoro-1,3-benzodioxole
SMILES C1=CC2=C(C=C1Br)OC(O2)(F)F

Specifications

Density 1.74
Boiling Point 78°C to 79°C (20 mmHg)
Refractive Index 1.4967
Quantity 5 g
Beilstein 1425209
Solubility Information Miscible with hexane.
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 5-Bromo-2,2-difluoro-1,3-benzodioxole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.