missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-dihydro-1,4-benzodioxane, 96%, BLD Pharm™

Product Code. 31110623
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
10 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31110623 5 g
31094926 250 mg
31108823 10 g
31092647 1 g
31092462 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31110623 Supplier BLD Pharm Supplier No. BD1026115g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 58328-39-5
Molecular Formula C8H7BrO2
MDL Number MFCD03095029
Quantity 5 g
SMILES BrC1=C2OCCOC2=CC=C1
Molecular Weight (g/mol) 215.04
Percent Purity 96%
Chemical Name or Material 5-Bromo-2,3-dihydro-1,4-benzodioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.