missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-dihydrobenzo[b]furan-7-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10432242
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432242 1 g Amber Glass Bottle 1g
10718065 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10432242 Supplier Thermo Scientific Maybridge Supplier No. MO07139DA
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 41177-72-4
Molecular Formula C9H7BrO3
Molecular Weight (g/mol) 243.06
MDL Number MFCD00191391
InChI Key LEBMKAXASFPSFA-UHFFFAOYSA-N
Synonym 5-bromo-2,3-dihydrobenzofuran-7-carboxylic acid, 5-bromo-2,3-dihydrobenzo b furan-7-carboxylic acid, 5-bromocoumaran-7-carboxylic acid, 5-bromo-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylic acid, 5-bromo-2,3-dihydro, zlchem 1161, acmc-1aqu8, 5-bromo-2,3-dihydrobenzofuran-7-carboxylicacid, 5-bromanyl-2,3-dihydro-1-benzofuran-7-carboxylic acid
PubChem CID 2794797
IUPAC Name 5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid
SMILES OC(=O)C1=CC(Br)=CC2=C1OCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 243.06
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 3-dihydrobenzo[b]furan-7-carboxylic acid, May contain varying amounts of anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.