missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dimethyl-1,3-thiazole, 97%, Thermo Scientific™

Product Code. 10650903
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10650903 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10650903 Supplier Thermo Scientific Maybridge Supplier No. CC05810DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 28599-52-2
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H6BrNS
Quantity 1 g
Synonym 5-bromo-2,4-dimethylthiazole, thiazole, 5-bromo-2,4-dimethyl, zlchem 569, pubchem9878, 2,4-dimethyl-5-bromothiazole, 5-bromo-2,4-dimethyl-thiazole, thiazole,5-bromo-2,4-dimethyl, 5-bromo-2,4-dimethyl-1,3thiazole
InChI Key BSFCVAYFJQLZEU-UHFFFAOYSA-N
SMILES CC1=C(SC(=N1)C)Br
IUPAC Name 5-bromo-2,4-dimethyl-1,3-thiazole
Molecular Weight (g/mol) 192.074
PubChem CID 298628
Formula Weight 192.08
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 4-dimethyl-1, 3-thiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.