missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-anisaldehyde, 99%, Thermo Scientific Chemicals

Catalog Number p-3756966
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
25016-01-7
C8H7BrO2
215.05
MFCD00003347
IJIBRSFAXRFPPN-UHFFFAOYSA-N
5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
90684
5-bromo-2-methoxybenzaldehyde
COC1=C(C=C(C=C1)Br)C=O
This item is not returnable. View return policy
25016-01-7
C8H7BrO2
215.05
MFCD00003347
IJIBRSFAXRFPPN-UHFFFAOYSA-N
5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
90684
5-bromo-2-methoxybenzaldehyde
COC1=C(C=C(C=C1)Br)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 118°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(OCH3)CHO
Quantity 5 g
Beilstein 08, 55
Formula Weight 215.05
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-anisaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-anisaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'