missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzoic Acid, TRC

Product Code. 30346052
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346052 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346052 Supplier Toronto Research Chemicals Supplier No. TRCB68212025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Bromo-2-chlorobenzoic Acid
CAS 21739-92-4
Molecular Formula C7H4BrClO2
Quantity 25 g
Synonym 2-Chloro-5-bromobenzoic acid
InChI Formula InChI=1S/C7H4BrClO2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,(H,10,11)
SMILES OC(=O)c1cc(Br)ccc1Cl
IUPAC Name 5-bromo-2-chlorobenzoic acid
Molecular Weight (g/mol) 235.46
Formula Weight 233.9083
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.