missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloropyridine, 98%, Thermo Scientific™

Product Code. 10018311 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10018311 1 g Glass bottle 1g
10186763 5 g Glass bottle 5g
10196763 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10018311 Supplier Thermo Scientific Supplier No. 360670010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 53939-30-3
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD01318951
InChI Key PEAOEIWYQVXZMB-UHFFFAOYSA-N
Synonym 2-chloro-5-bromopyridine, 5-bromo-2-chloro-pyridine, pyridine, 5-bromo-2-chloro, 2-chloro-5-bromo-pyridine, 2-chlorine-5-bromine pyridine, pubchem1168, acmc-1afhf, 3-bromo-6-chloropyridine, 2-chloro-5-bromo pyridine, 3-bromo-6-chloro-pyridine
PubChem CID 2734414
IUPAC Name 5-bromo-2-chloropyridine
SMILES C1=CC(=NC=C1Br)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 67°C to 71°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 192.44
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 5-Bromo-2-chloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-chloropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.