missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloropyrimidine, 98+%, Thermo Scientific Chemicals

Product Code. 15469079
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469079 25 g 25g
11384975 1 g 1g
11394975 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15469079 Supplier Thermo Scientific Alfa Aesar Supplier No. L18360.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32779-36-5 | Molecular Formula: C4H2BrClN2 | Molecular Weight: 193.43 g/mol

It is applied as a pharmaceutical intermediate, in the synthesis of pharmaceutical goods such as inhibitors. It is also employed in the cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a pharmaceutical intermediate, in the synthesis of pharmaceutical goods such as inhibitors. It is also employed in the cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.

Solubility
Insoluble in water. Slightly soluble in DMSO and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32779-36-5
Molecular Formula C4H2BrClN2
Molecular Weight (g/mol) 193.43
MDL Number MFCD00483232
InChI Key XPGIBDJXEVAVTO-UHFFFAOYSA-N
Synonym 2-chloro-5-bromopyrimidine, 5-bromo-2-chloro-pyrimidine, 5-bromo-2-chloropyrimidne, pyrimidine, 5-bromo-2-chloro, 5-bromo-2-chloro-1,3-diazine, pubchem5159, zlchem 1210, acmc-209hvm, 5-bromo chloropyrimidine, 5-bromo-2-chloroprimidine
PubChem CID 606665
IUPAC Name 5-bromo-2-chloropyrimidine
SMILES ClC1=NC=C(Br)C=N1

Specifications

Melting Point 77°C to 80°C
Boiling Point 95°C (15 mmHg)
Quantity 25 g
Beilstein 507955
Solubility Information Insoluble aq. soln.. Slightly soluble in DMSO and methanol.
Formula Weight 193.43
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-chloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.