missing translation for 'onlineSavingsMsg'
Learn More

Melford 5-Bromo-2'-Deoxyuridine

Product Code. p-200087028
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 30018122

Brand: Melford B742000.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thymidine analog used for study of DNA synthesis.

Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.

Specifications

CAS 59-14-3
Molecular Formula C9H11BrN2O5
Quantity 500 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.