missing translation for 'onlineSavingsMsg'
Learn More

Melford 5-Bromo-2'-Deoxyuridine

Product Code. 30018124
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018124 5 g 5g
30018122 500 mg 500mg
30018123 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018124 Supplier Melford Supplier No. B742005.0
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thymidine analog used for study of DNA synthesis.

Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.

Specifications

CAS 59-14-3
Molecular Formula C9H11BrN2O5
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.