missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2'-deoxyuridine, Fisher BioReagents

Product Code. 11453064
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Amber Glass
Unit Size:
1g
250mg
500mg
5g
Product Code. Quantity Packaging unitSize
11453064 1 g Amber Glass 1g
10606005 250 mg Amber Glass 250mg
10328514 500 mg Amber Glass 500mg
10092604 5 g Amber Glass 5g
4 options
This item is not returnable. View return policy

Product Code. 11453064

Brand: Fisher BioReagents BP25081

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
5-Bromo-2'-Deoxyuridine is a thymidine analog used as a mutagen in genetic research.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-14-3
Molecular Formula C9H11BrN2O5
Molecular Weight (g/mol) 307.10
MDL Number MFCD00006529
InChI Key WOVKYSAHUYNSMH-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
PubChem CID 6035
ChEBI CHEBI:472552
IUPAC Name 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O

Specifications

Melting Point 191°C
Color White
Infrared Spectrum Conforms to reference
Assay Percent Range >99%
Packaging Amber Glass
Quantity 1 g
Merck Index 15, 1461
Formula Weight 307.1
Physical Form Powder or Solid
Chemical Name or Material 5-Bromo-2'-Deoxyuridine, Off-white Powder
compliance-icons compliance-icons
Product Identifier
  • 5-Bromo-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.