missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(difluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

Product Code. 15412729
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412729 1 g 1g
15422729 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15412729 Supplier Thermo Scientific Alfa Aesar Supplier No. H66297.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845827-13-6
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD11977429
InChI Key QXLZRIGSWWQOLG-UHFFFAOYSA-N
Synonym 5-bromo-2-difluoromethyl pyridine, pyridine, 5-bromo-2-difluoromethyl, 2-difluoromethyl-5-bromopyridine, pyridine,5-bromo-2-difluoromethyl, 3-bromo-6-difluoromethyl pyridine, 2-bis fluoranyl methyl-5-bromanyl-pyridine, 5-bromo-alpha,alpha-difluoro-2-picoline
PubChem CID 53415062
IUPAC Name 5-bromo-2-(difluoromethyl)pyridine
SMILES FC(F)C1=NC=C(Br)C=C1

Specifications

Melting Point 35°C to 40°C
Color White to Yellow
Flash Point 89°C (192°F)
Quantity 1 g
UN Number UN2811
Formula Weight 208
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-(difluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.