missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoropyridine, 99%

Product Code. 10095782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10095782 5 g Glass bottle 5g
10005792 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10095782 Supplier Thermo Scientific Acros Supplier No. 378460050
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 766-11-0 | C5H3BrFN | 175.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-11-0
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.99
MDL Number MFCD01863742
InChI Key MYUQKYGWKHTRPG-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromopyridine, 3-bromo-6-fluoropyridine, 5-bromo-2-fluoro pyridine, 5-bromo-2-fluoro-pyridine, pyridine, 5-bromo-2-fluoro, 2-fluoro-5-bromo pyridine, pubchem3550, 2-fluro-5-bromo-pyridine, acmc-209p4s, bromo-2-fluoropyridine, 5
PubChem CID 2783168
IUPAC Name 5-bromo-2-fluoropyridine
SMILES C1=CC(=NC=C1Br)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.7200g/mL
Boiling Point 170.0°C to 173.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5310 to 1.5330
Quantity 5 g
Specific Gravity 1.72
Formula Weight 175.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluoropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-fluoropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.