missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxy-3-nitrobenzaldehyde, 97%, BLD Pharm™

Product Code. 30803858
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30803858 5 g 5g
30796693 100 g 100g
30802285 1 g 1g
30794141 250 mg 250mg
30786780 25 g 25g
30787715 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30803858 Supplier BLD Pharm Supplier No. BD1029075g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16634-88-1
Molecular Formula C7H4BrNO4
MDL Number MFCD00130110
Quantity 5 g
SMILES OC1=C(C=O)C=C(Br)C=C1[N+]([O-])=O
Molecular Weight (g/mol) 246.02
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxy-3-nitrobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.